The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005891.2 | NP_005882.2 | 1 - 2 | 193 | 80 |
GenBank mRNA | AB208993.1 | - | 1 - 2 | 83 | 80 |
AF356877.1 | - | 1 - 2 | 116 | 80 | |
AK225089.1 | - | 1 - 2 | 94 | 80 | |
AK225244.1 | - | 1 - 2 | 106 | 80 | |
AK291080.1 | - | 1 - 2 | 94 | 80 | |
BC000408.2 | - | 1 - 2 | 73 | 80 | |
BP210609.1 | - | 1 - 2 | 193 | 80 | |
S70154.1 | - | 1 - 2 | 99 | 80 |