The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018992.3 | NP_061865.1 | 3 - 4 | 506 | 124 |
GenBank mRNA | AK000047.1 | - | 3 - 4 | 504 | 124 |
BC007314.2 | - | 3 - 4 | 459 | 124 | |
DA134449.1 | - | 3 - 4 | 506 | 124 |