Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARP4
Entrez Gene ID
143
Gene Name
poly(ADP-ribose) polymerase family member 4
Gene Aliases
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193
Chromosome Location
Chr.13: 24420930 - 24512993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006437.3 NP_006428.2 32 - 33 4953 126
XM_011534931.1 XP_011533233.1 31 - 32 5003 126
XM_011534932.2 XP_011533234.1 32 - 33 5170 126
GenBank mRNA AF057160.1 - 32 - 33 4899 126
AF158255.1 - 32 - 33 4953 126
CA308288.1 - 2 - 3 126
D79999.2 - 31 - 32 4852 126

View Full Product Details