The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006437.3 | NP_006428.2 | 4 - 5 | 500 | 120 |
XM_011534931.1 | XP_011533233.1 | 3 - 4 | 550 | 120 | |
XM_011534932.2 | XP_011533234.1 | 4 - 5 | 717 | 120 | |
GenBank mRNA | AF057160.1 | - | 4 - 5 | 446 | 120 |
AF158255.1 | - | 4 - 5 | 500 | 120 | |
AK304399.1 | - | 4 - 5 | 468 | 120 | |
BC017815.1 | - | 4 - 5 | 439 | 120 | |
D79999.2 | - | 3 - 4 | 399 | 120 | |
DB238428.1 | - | 4 - 5 | 468 | 120 |