The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010875.3 | NP_001010875.1 | 7 - 8 | 827 | 94 |
NM_001286806.1 | NP_001273735.1 | 6 - 7 | 708 | 94 | |
NM_001286807.1 | NP_001273736.1 | 7 - 8 | 885 | 94 | |
XM_005266321.2 | XP_005266378.1 | 7 - 8 | 753 | 94 | |
XM_005266322.1 | XP_005266379.1 | 7 - 8 | 753 | 94 | |
XM_006719793.3 | XP_006719856.1 | 7 - 8 | 999 | 94 | |
XM_006719794.1 | XP_006719857.1 | 7 - 8 | 877 | 94 | |
XM_011535026.1 | XP_011533328.1 | 6 - 7 | 604 | 94 | |
XM_011535027.2 | XP_011533329.1 | 6 - 7 | 604 | 94 | |
XM_017020523.1 | XP_016876012.1 | 7 - 8 | 824 | 94 | |
XM_017020524.1 | XP_016876013.1 | 7 - 8 | 877 | 94 | |
GenBank mRNA | AK094133.1 | - | 7 - 8 | 754 | 94 |
AK095465.1 | - | 6 - 7 | 607 | 94 | |
AL832206.1 | - | 3 - 4 | 280 | 94 | |
BC132739.1 | - | 7 - 8 | 750 | 94 | |
BC136760.1 | - | 7 - 8 | 750 | 94 | |
BC144401.1 | - | 7 - 8 | 808 | 94 |