The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007118.3 | NP_009049.2 | 18 - 19 | 3245 | 56 |
NR_134469.1 | 18 - 19 | 3245 | 56 | ||
XM_011514107.2 | XP_011512409.1 | 18 - 19 | 3820 | 56 | |
XM_011514108.1 | XP_011512410.1 | 18 - 19 | 3119 | 56 | |
XM_011514109.2 | XP_011512411.1 | 18 - 19 | 6897 | 56 | |
XM_011514110.2 | XP_011512412.1 | 18 - 19 | 3355 | 56 | |
XM_017009801.1 | XP_016865290.1 | 18 - 19 | 3597 | 56 | |
XM_017009802.1 | XP_016865291.1 | 18 - 19 | 3597 | 56 | |
XM_017009803.1 | XP_016865292.1 | 11 - 12 | 1866 | 56 | |
GenBank mRNA | AB209754.1 | - | 13 - 14 | 2139 | 56 |
AF091395.1 | - | 18 - 19 | 3110 | 56 | |
AK304786.1 | - | 18 - 19 | 3224 | 56 | |
BC035585.1 | - | 18 - 19 | 3170 | 56 | |
U42390.1 | - | 18 - 19 | 3110 | 56 |