The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007124.2 | NP_009055.2 | 52 - 53 | 7907 | 83 |
XM_005267127.4 | XP_005267184.1 | 52 - 53 | 8242 | 83 | |
XM_005267130.2 | XP_005267187.1 | 53 - 54 | 8125 | 83 | |
XM_005267133.2 | XP_005267190.1 | 52 - 53 | 8925 | 83 | |
XM_006715560.3 | XP_006715623.1 | 3 - 4 | 1249 | 83 | |
XM_011536101.2 | XP_011534403.1 | 54 - 55 | 8586 | 83 | |
XM_011536102.2 | XP_011534404.1 | 54 - 55 | 8127 | 83 | |
XM_011536106.2 | XP_011534408.1 | 53 - 54 | 8774 | 83 | |
XM_011536107.2 | XP_011534409.1 | 53 - 54 | 8150 | 83 | |
XM_011536109.2 | XP_011534411.1 | 3 - 4 | 2556 | 83 | |
XM_017011243.1 | XP_016866732.1 | 54 - 55 | 8124 | 83 | |
XM_017011244.1 | XP_016866733.1 | 54 - 55 | 8127 | 83 | |
GenBank mRNA | X69086.1 | - | 52 - 53 | 7815 | 83 |