Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTRN
Entrez Gene ID
7402
Gene Name
utrophin
Gene Aliases
DMDL, DRP, DRP1
Chromosome Location
Chr.6: 144284955 - 144853034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007124.2 NP_009055.2 71 - 72 10268 86
XM_005267127.4 XP_005267184.1 71 - 72 10603 86
XM_005267130.2 XP_005267187.1 72 - 73 10486 86
XM_005267133.2 XP_005267190.1 71 - 72 11286 86
XM_006715560.3 XP_006715623.1 22 - 23 3610 86
XM_011536101.2 XP_011534403.1 73 - 74 10947 86
XM_011536102.2 XP_011534404.1 73 - 74 10488 86
XM_011536106.2 XP_011534408.1 72 - 73 11135 86
XM_011536107.2 XP_011534409.1 72 - 73 10511 86
XM_011536109.2 XP_011534411.1 22 - 23 4917 86
XM_017011243.1 XP_016866732.1 73 - 74 10485 86
XM_017011244.1 XP_016866733.1 72 - 73 10449 86
GenBank mRNA BX100445.1 - 2 - 3 161 86
X15488.1 - 11 - 12 1347 86
X69086.1 - 71 - 72 10176 86

View Full Product Details