Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTRN
Entrez Gene ID
7402
Gene Name
utrophin
Gene Aliases
DMDL, DRP, DRP1
Chromosome Location
Chr.6: 144284955 - 144853034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007124.2 NP_009055.2 73 - 74 10387 65
XM_005267127.4 XP_005267184.1 73 - 74 10722 65
XM_005267130.2 XP_005267187.1 74 - 75 10605 65
XM_005267133.2 XP_005267190.1 73 - 74 11405 65
XM_006715560.3 XP_006715623.1 24 - 25 3729 65
XM_011536101.2 XP_011534403.1 75 - 76 11066 65
XM_011536102.2 XP_011534404.1 75 - 76 10607 65
XM_011536106.2 XP_011534408.1 74 - 75 11254 65
XM_011536107.2 XP_011534409.1 74 - 75 10630 65
XM_011536109.2 XP_011534411.1 24 - 25 5036 65
XM_017011243.1 XP_016866732.1 75 - 76 10604 65
XM_017011244.1 XP_016866733.1 74 - 75 10568 65
GenBank mRNA BX100445.1 - 4 - 5 280 65

View Full Product Details