Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA0141
Entrez Gene ID
9812
Gene Name
KIAA0141
Gene Aliases
DELE
Chromosome Location
Chr.5: 141923785 - 141942047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142603.2 NP_001136075.1 7 - 8 878 104
NM_014773.4 NP_055588.3 7 - 8 878 104
XM_005268547.3 XP_005268604.1 7 - 8 913 104
XM_005268548.3 XP_005268605.1 7 - 8 913 104
XM_005268552.2 XP_005268609.1 7 - 8 913 104
XM_011537715.1 XP_011536017.1 7 - 8 913 104
GenBank mRNA AK292869.1 - 7 - 8 827 104
BC007855.2 - 7 - 8 854 104
BC010452.1 - 7 - 8 769 104
BC011269.1 - 7 - 8 769 104
BC011926.1 - 7 - 8 799 104
BC035486.1 - 7 - 8 794 104
D50931.1 - 7 - 8 836 104

View Full Product Details