The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135862.2 | NP_001129334.1 | 11 - 12 | 1180 | 53 |
NM_001242450.1 | NP_001229379.1 | 10 - 11 | 1220 | 53 | |
NM_001284296.1 | NP_001271225.1 | 11 - 12 | 1266 | 53 | |
NM_138415.4 | NP_612424.1 | 10 - 11 | 1351 | 53 | |
NR_134535.1 | 9 - 10 | 1112 | 53 | ||
XM_006724122.1 | XP_006724185.1 | 10 - 11 | 1360 | 53 | |
GenBank mRNA | AK091480.1 | - | 11 - 12 | 1266 | 53 |
AK092243.1 | - | 11 - 12 | 1180 | 53 | |
AK095777.1 | - | 4 - 5 | 332 | 53 | |
AK297317.1 | - | 10 - 11 | 1213 | 53 | |
BC012187.1 | - | 10 - 11 | 1351 | 53 | |
CR456448.1 | - | 10 - 11 | 1107 | 53 | |
KU178842.1 | - | 10 - 11 | 1200 | 53 | |
KU178843.1 | - | 10 - 11 | 1074 | 53 | |
KU178844.1 | - | 9 - 10 | 1051 | 53 |