The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000602.4 | NP_000593.1 | 1 - 2 | 159 | 54 |
XM_017012260.1 | XP_016867749.1 | 1 - 2 | 161 | 54 | |
GenBank mRNA | AK297728.1 | - | 1 - 2 | 146 | 54 |
AK316225.1 | - | 1 - 2 | 145 | 54 | |
BC010860.1 | - | 1 - 2 | 124 | 54 | |
CN309001.1 | - | 1 - 2 | 160 | 54 | |
M16006.1 | - | 1 - 2 | 77 | 54 | |
X04429.1 | - | 1 - 2 | 128 | 54 | |
X04729.1 | - | 1 - 2 | 145 | 54 | |
X12701.1 | - | 1 - 2 | 128 | 54 |