Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
Chromosome Location
Chr.15: 49858238 - 50182096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024837.3 NP_079113.2 12 - 13 1170 145
NR_073596.1 13 - 14 1258 145
NR_073597.1 12 - 13 1170 145
NR_073598.1 13 - 14 1277 145
XM_011522046.2 XP_011520348.1 14 - 15 1559 145
XM_011522047.2 XP_011520349.1 13 - 14 2084 145
XM_011522048.1 XP_011520350.1 13 - 14 1357 145
XM_011522049.2 XP_011520351.1 13 - 14 1467 145
XM_011522051.2 XP_011520353.1 14 - 15 1394 145
XM_011522052.2 XP_011520354.1 13 - 14 1452 145
XM_011522053.1 XP_011520355.1 13 - 14 1233 145
XM_011522056.2 XP_011520358.2 13 - 14 1619 145
XM_011522058.2 XP_011520360.1 9 - 10 1828 145
XM_011522059.1 XP_011520361.1 8 - 9 849 145
XM_011522060.1 XP_011520362.1 11 - 12 1084 145
XM_011522061.1 XP_011520363.1 10 - 11 999 145
XM_011522062.1 XP_011520364.1 8 - 9 838 145
XM_011522063.1 XP_011520365.1 8 - 9 820 145
XM_011522064.1 XP_011520366.1 4 - 5 521 145
XM_011522069.2 XP_011520371.1 3 - 4 342 145
XM_017022587.1 XP_016878076.1 12 - 13 1535 145
XM_017022588.1 XP_016878077.1 12 - 13 1460 145
XM_017022589.1 XP_016878078.1 13 - 14 1619 145
XM_017022590.1 XP_016878079.1 14 - 15 1371 145
XM_017022591.1 XP_016878080.1 13 - 14 1318 145
XM_017022592.1 XP_016878081.1 12 - 13 1381 145
XM_017022593.1 XP_016878082.1 13 - 14 1619 145
XM_017022594.1 XP_016878083.1 9 - 10 879 145
XM_017022595.1 XP_016878084.1 9 - 10 2884 145
XM_017022596.1 XP_016878085.1 4 - 5 512 145
XM_017022597.1 XP_016878086.1 13 - 14 1619 145
XM_017022598.1 XP_016878087.1 13 - 14 1619 145
XM_017022599.1 XP_016878088.1 13 - 14 1619 145
GenBank mRNA AB075819.1 - 13 - 14 1277 145
BC144297.1 - 12 - 13 1166 145

View Full Product Details