Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
Chromosome Location
Chr.15: 49858238 - 50182096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024837.3 NP_079113.2 19 - 20 2182 143
NR_073596.1 19 - 20 2081 143
NR_073597.1 19 - 20 2182 143
NR_073598.1 20 - 21 2289 143
XM_011522046.2 XP_011520348.1 21 - 22 2571 143
XM_011522047.2 XP_011520349.1 20 - 21 3096 143
XM_011522048.1 XP_011520350.1 20 - 21 2369 143
XM_011522049.2 XP_011520351.1 20 - 21 2479 143
XM_011522051.2 XP_011520353.1 21 - 22 2406 143
XM_011522052.2 XP_011520354.1 20 - 21 2464 143
XM_011522053.1 XP_011520355.1 20 - 21 2245 143
XM_011522056.2 XP_011520358.2 20 - 21 2631 143
XM_011522058.2 XP_011520360.1 16 - 17 2840 143
XM_011522059.1 XP_011520361.1 15 - 16 1861 143
XM_011522060.1 XP_011520362.1 18 - 19 2096 143
XM_011522061.1 XP_011520363.1 17 - 18 2011 143
XM_011522062.1 XP_011520364.1 15 - 16 1850 143
XM_011522063.1 XP_011520365.1 15 - 16 1832 143
XM_011522064.1 XP_011520366.1 11 - 12 1533 143
XM_011522069.2 XP_011520371.1 10 - 11 1354 143
XM_011522070.1 XP_011520372.1 6 - 7 774 143
XM_017022587.1 XP_016878076.1 19 - 20 2547 143
XM_017022588.1 XP_016878077.1 19 - 20 2472 143
XM_017022589.1 XP_016878078.1 19 - 20 2442 143
XM_017022590.1 XP_016878079.1 21 - 22 2383 143
XM_017022591.1 XP_016878080.1 20 - 21 2330 143
XM_017022592.1 XP_016878081.1 19 - 20 2393 143
XM_017022593.1 XP_016878082.1 20 - 21 2631 143
XM_017022594.1 XP_016878083.1 16 - 17 1891 143
XM_017022595.1 XP_016878084.1 16 - 17 3896 143
XM_017022596.1 XP_016878085.1 11 - 12 1524 143
GenBank mRNA AB075819.1 - 20 - 21 2289 143
BC144297.1 - 19 - 20 2178 143
BF898163.1 - 1 - 2 143

View Full Product Details