Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
Chromosome Location
Chr.15: 49858238 - 50182096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024837.3 NP_079113.2 10 - 11 891 192
NR_073596.1 11 - 12 979 192
NR_073597.1 10 - 11 891 192
NR_073598.1 11 - 12 998 192
XM_011522046.2 XP_011520348.1 12 - 13 1280 192
XM_011522047.2 XP_011520349.1 11 - 12 1805 192
XM_011522048.1 XP_011520350.1 11 - 12 1078 192
XM_011522049.2 XP_011520351.1 11 - 12 1188 192
XM_011522051.2 XP_011520353.1 12 - 13 1115 192
XM_011522052.2 XP_011520354.1 11 - 12 1173 192
XM_011522053.1 XP_011520355.1 11 - 12 954 192
XM_011522056.2 XP_011520358.2 11 - 12 1340 192
XM_011522058.2 XP_011520360.1 7 - 8 1549 192
XM_011522059.1 XP_011520361.1 6 - 7 570 192
XM_011522060.1 XP_011520362.1 9 - 10 805 192
XM_011522061.1 XP_011520363.1 8 - 9 720 192
XM_011522062.1 XP_011520364.1 6 - 7 559 192
XM_011522063.1 XP_011520365.1 6 - 7 541 192
XM_011522064.1 XP_011520366.1 2 - 3 242 192
XM_017022587.1 XP_016878076.1 10 - 11 1256 192
XM_017022589.1 XP_016878078.1 11 - 12 1340 192
XM_017022590.1 XP_016878079.1 12 - 13 1092 192
XM_017022591.1 XP_016878080.1 11 - 12 1039 192
XM_017022592.1 XP_016878081.1 10 - 11 1102 192
XM_017022593.1 XP_016878082.1 11 - 12 1340 192
XM_017022594.1 XP_016878083.1 7 - 8 600 192
XM_017022595.1 XP_016878084.1 7 - 8 2605 192
XM_017022596.1 XP_016878085.1 2 - 3 233 192
XM_017022597.1 XP_016878086.1 11 - 12 1340 192
XM_017022598.1 XP_016878087.1 11 - 12 1340 192
XM_017022599.1 XP_016878088.1 11 - 12 1340 192
GenBank mRNA AB075819.1 - 11 - 12 998 192
BC144297.1 - 10 - 11 887 192

View Full Product Details