The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286430.1 | NP_001273359.1 | 6 - 7 | 1345 | 63 |
NM_001323903.1 | NP_001310832.1 | 8 - 9 | 1525 | 63 | |
NM_001323904.1 | NP_001310833.1 | 8 - 9 | 1525 | 63 | |
NM_138792.3 | NP_620147.1 | 8 - 9 | 1525 | 63 | |
XM_017021911.1 | XP_016877400.1 | 8 - 9 | 1522 | 63 | |
GenBank mRNA | AK055762.1 | - | 6 - 7 | 1321 | 63 |
AK297972.1 | - | 9 - 10 | 1435 | 63 | |
AY302186.1 | - | 8 - 9 | 1509 | 63 | |
BC018147.2 | - | 8 - 9 | 1485 | 63 |