The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002223.3 | NP_002214.2 | 44 - 45 | 6635 | 106 |
XM_017019266.1 | XP_016874755.1 | 45 - 46 | 6694 | 106 | |
XM_017019267.1 | XP_016874756.1 | 45 - 46 | 6218 | 106 | |
XM_017019268.1 | XP_016874757.1 | 44 - 45 | 6095 | 106 | |
GenBank mRNA | AK307686.1 | - | 7 - 8 | 1046 | 106 |
D26350.1 | - | 44 - 45 | 6636 | 106 |