The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002223.3 | NP_002214.2 | 51 - 52 | 7722 | 154 |
XM_017019266.1 | XP_016874755.1 | 52 - 53 | 7781 | 154 | |
XM_017019267.1 | XP_016874756.1 | 52 - 53 | 7305 | 154 | |
XM_017019268.1 | XP_016874757.1 | 51 - 52 | 7182 | 154 | |
GenBank mRNA | AK307686.1 | - | 14 - 15 | 2133 | 154 |
D26350.1 | - | 51 - 52 | 7723 | 154 |