The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170753.1 | NP_001164224.1 | 3 - 4 | 362 | 69 |
NM_145017.2 | NP_659454.2 | 3 - 4 | 362 | 69 | |
GenBank mRNA | AK057333.1 | - | 3 - 4 | 355 | 69 |
BC043246.1 | - | 3 - 4 | 360 | 69 | |
BC053995.1 | - | 2 - 3 | 259 | 69 | |
DB047394.1 | - | 3 - 4 | 362 | 69 | |
JF442033.1 | - | 3 - 4 | 335 | 69 |