Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFP
Entrez Gene ID
10186
Gene Name
lipoma HMGIC fusion partner
Gene Aliases
-
Chromosome Location
Chr.13: 39342892 - 39603219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005780.2 NP_005771.1 1 - 2 338 73
GenBank mRNA AF098807.1 - 1 - 2 183 73
AK129819.1 - 1 - 2 290 73
AK223369.1 - 1 - 2 290 73
BC017824.1 - 1 - 2 186 73
DA375214.1 - 1 - 2 338 73

View Full Product Details