Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROCK1
Entrez Gene ID
6093
Gene Name
Rho associated coiled-coil containing protein kinase 1
Gene Aliases
P160ROCK, ROCK-I
Chromosome Location
Chr.18: 20949740 - 21111851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005406.2 NP_005397.1 17 - 18 2927 137
XM_011526137.2 XP_011524439.1 6 - 7 1161 137
GenBank mRNA AB208965.1 - 9 - 10 1020 137
AK307388.1 - 17 - 18 2742 137
BC113114.1 - 17 - 18 2002 137
U43195.1 - 17 - 18 1986 137

View Full Product Details