Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD2
Entrez Gene ID
5134
Gene Name
programmed cell death 2
Gene Aliases
RP8, ZMYND7
Chromosome Location
Chr.6: 170575572 - 170584692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017010926.1 XP_016866415.1 5 - 6 926 75

View Full Product Details