The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002793.3 | NP_002784.1 | 1 - 2 | 194 | 132 |
GenBank mRNA | AB209078.1 | - | 1 - 2 | 124 | 132 |
AB451312.1 | - | 1 - 2 | 107 | 132 | |
AB451442.1 | - | 1 - 2 | 107 | 132 | |
AK058050.1 | - | 1 - 2 | 164 | 132 | |
AK223193.1 | - | 1 - 2 | 145 | 132 | |
BC000508.2 | - | 1 - 2 | 145 | 132 | |
BC020807.1 | - | 1 - 2 | 145 | 132 | |
BP369175.1 | - | 1 - 2 | 206 | 132 | |
BT019720.1 | - | 1 - 2 | 107 | 132 | |
D00761.1 | - | 1 - 2 | 119 | 132 |