The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020732.3 | NP_065783.3 | 2 - 3 | 1736 | 78 |
XM_011535984.2 | XP_011534286.2 | 2 - 3 | 1812 | 78 | |
XM_017011103.1 | XP_016866592.1 | 2 - 3 | 1812 | 78 | |
XM_017011104.1 | XP_016866593.1 | 2 - 3 | 1811 | 78 | |
XM_017011105.1 | XP_016866594.1 | 2 - 3 | 1812 | 78 | |
XM_017011106.1 | XP_016866595.1 | 2 - 3 | 1811 | 78 | |
XM_017011107.1 | XP_016866596.1 | 2 - 3 | 1812 | 78 | |
XM_017011108.1 | XP_016866597.1 | 2 - 3 | 1819 | 78 | |
GenBank mRNA | AF468300.1 | - | 2 - 3 | 266 | 78 |