The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017519.2 | NP_059989.2 | 7 - 8 | 2508 | 124 |
NM_020732.3 | NP_065783.3 | 8 - 9 | 2547 | 124 | |
XM_005267069.4 | XP_005267126.2 | 7 - 8 | 2583 | 124 | |
XM_011535984.2 | XP_011534286.2 | 9 - 10 | 2665 | 124 | |
XM_017011103.1 | XP_016866592.1 | 9 - 10 | 2665 | 124 | |
XM_017011104.1 | XP_016866593.1 | 9 - 10 | 2664 | 124 | |
XM_017011105.1 | XP_016866594.1 | 9 - 10 | 2665 | 124 | |
XM_017011106.1 | XP_016866595.1 | 9 - 10 | 2664 | 124 | |
XM_017011108.1 | XP_016866597.1 | 9 - 10 | 2672 | 124 | |
XM_017011109.1 | XP_016866598.1 | 5 - 6 | 5021 | 124 | |
GenBank mRNA | AB033061.1 | - | 1 - 2 | 96 | 124 |
AF259792.1 | - | 3 - 4 | 450 | 124 | |
AF468300.1 | - | 8 - 9 | 1077 | 124 | |
AF521671.1 | - | 7 - 8 | 2295 | 124 | |
AK025945.1 | - | 5 - 6 | 774 | 124 | |
AK074180.1 | - | 5 - 6 | 658 | 124 |