Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID1B
Entrez Gene ID
57492
Gene Name
AT-rich interaction domain 1B
Gene Aliases
6A3-5, BAF250B, BRIGHT, CSS1, DAN15, ELD/OSA1, MRD12, OSA2, P250R
Chromosome Location
Chr.6: 156777847 - 157210779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005267069.4 XP_005267126.2 11 - 12 3330 69
XM_011535984.2 XP_011534286.2 14 - 15 3541 69
XM_011535988.2 XP_011534290.1 4 - 5 484 69
XM_017011103.1 XP_016866592.1 13 - 14 3442 69
XM_017011104.1 XP_016866593.1 13 - 14 3411 69
XM_017011107.1 XP_016866596.1 12 - 13 3232 69
XM_017011108.1 XP_016866597.1 14 - 15 3548 69
XM_017011109.1 XP_016866598.1 9 - 10 5768 69
GenBank mRNA AB033061.1 - 5 - 6 843 69
AK025945.1 - 9 - 10 1521 69

View Full Product Details