The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146282.1 | NP_001139754.1 | 4 - 5 | 611 | 73 |
NM_003136.3 | NP_003127.1 | 5 - 6 | 703 | 73 | |
XM_005268024.2 | XP_005268081.1 | 5 - 6 | 956 | 73 | |
XM_011537106.1 | XP_011535408.1 | 5 - 6 | 584 | 73 | |
XM_017021615.1 | XP_016877104.1 | 4 - 5 | 543 | 73 | |
GenBank mRNA | AK293319.1 | - | 5 - 6 | 495 | 73 |
AK300824.1 | - | 4 - 5 | 484 | 73 | |
AK312853.1 | - | 5 - 6 | 648 | 73 | |
BC000652.2 | - | 5 - 6 | 570 | 73 | |
BC003389.1 | - | 5 - 6 | 576 | 73 | |
U51920.1 | - | 5 - 6 | 423 | 73 |