The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001137673.1 | NP_001131145.1 | 2 - 3 | 320 | 140 |
NM_004516.3 | NP_004507.2 | 2 - 3 | 320 | 140 | |
NM_012218.3 | NP_036350.2 | 2 - 3 | 320 | 140 | |
NM_017620.2 | NP_060090.2 | 2 - 3 | 320 | 140 | |
NM_153464.2 | NP_703194.1 | 2 - 3 | 320 | 140 | |
XM_006722742.2 | XP_006722805.1 | 2 - 3 | 235 | 140 | |
XM_011527985.2 | XP_011526287.1 | 2 - 3 | 235 | 140 | |
XM_017026765.1 | XP_016882254.1 | 2 - 3 | 235 | 140 | |
GenBank mRNA | AB209062.1 | - | 2 - 3 | 277 | 140 |
AF141870.1 | - | 2 - 3 | 114 | 140 | |
AK291205.1 | - | 2 - 3 | 219 | 140 | |
AK291617.1 | - | 2 - 3 | 205 | 140 | |
AK294084.1 | - | 2 - 3 | 211 | 140 | |
BC001770.1 | - | 2 - 3 | 205 | 140 | |
BC048314.1 | - | 2 - 3 | 250 | 132 | |
BC064836.1 | - | 2 - 3 | 253 | 140 | |
DC320902.1 | - | 2 - 3 | 323 | 140 | |
U10324.1 | - | 2 - 3 | 267 | 140 | |
X98264.1 | - | 2 - 3 | 207 | 140 |