The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004516.3 | NP_004507.2 | 13 - 14 | 1866 | 58 |
NM_012218.3 | NP_036350.2 | 13 - 14 | 1866 | 58 | |
NM_153464.2 | NP_703194.1 | 13 - 14 | 1866 | 58 | |
GenBank mRNA | AF141870.1 | - | 13 - 14 | 1660 | 58 |
AF147209.1 | - | 11 - 12 | 1561 | 58 | |
AF167569.1 | - | 11 - 12 | 1549 | 58 | |
AF167570.1 | - | 11 - 12 | 1549 | 58 | |
AJ271745.1 | - | 12 - 13 | 1638 | 58 | |
AJ271746.1 | - | 12 - 13 | 1618 | 58 | |
AK291205.1 | - | 13 - 14 | 1765 | 58 | |
AK294084.1 | - | 9 - 10 | 1169 | 58 | |
HQ322263.1 | - | 11 - 12 | 1549 | 58 | |
X98265.1 | - | 12 - 13 | 1758 | 58 |