Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr.10: 32446082 - 32882864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026383.2 NP_001021554.1 8 - 9 1099 75
NM_001321115.1 NP_001308044.1 8 - 9 1333 75
NM_145023.5 NP_659460.3 8 - 9 1333 75
NR_109826.1 8 - 9 1099 75
NR_109827.1 7 - 8 1166 75
XM_006717505.1 XP_006717568.1 8 - 9 1041 75
XM_006717506.1 XP_006717569.1 8 - 9 1041 75
XM_011519663.1 XP_011517965.1 8 - 9 1041 75
XM_011519664.1 XP_011517966.1 8 - 9 1041 75
XM_011519666.1 XP_011517968.1 8 - 9 1041 75
XM_011519667.1 XP_011517969.1 8 - 9 1041 75
XM_011519668.1 XP_011517970.1 8 - 9 1041 75
XM_011519669.1 XP_011517971.1 8 - 9 1041 75
XM_011519670.1 XP_011517972.1 8 - 9 1041 75
XM_011519671.1 XP_011517973.1 8 - 9 1041 75
XM_011519672.1 XP_011517974.1 8 - 9 1041 75
XM_011519673.1 XP_011517975.1 8 - 9 1041 75
XM_011519674.1 XP_011517976.1 8 - 9 1041 75
XM_011519675.1 XP_011517977.1 8 - 9 1041 75
XM_011519676.1 XP_011517978.1 8 - 9 1041 75
XM_011519677.1 XP_011517979.1 8 - 9 1041 75
XM_011519678.1 XP_011517980.1 8 - 9 1041 75
XM_011519679.1 XP_011517981.1 8 - 9 1041 75
XM_011519680.2 XP_011517982.1 3 - 4 176 75
XM_011519684.2 XP_011517986.1 8 - 9 1041 75
XM_011519685.1 XP_011517987.1 2 - 3 179 75
XM_011519686.2 XP_011517988.1 8 - 9 1041 75
XM_011519688.2 XP_011517990.1 8 - 9 1041 75
XM_017016638.1 XP_016872127.1 8 - 9 1041 75
XM_017016639.1 XP_016872128.1 8 - 9 1041 75
XM_017016640.1 XP_016872129.1 8 - 9 1041 75
XM_017016641.1 XP_016872130.1 3 - 4 470 75
XM_017016642.1 XP_016872131.1 3 - 4 543 75
XM_017016643.1 XP_016872132.1 3 - 4 160 75
XM_017016644.1 XP_016872133.1 8 - 9 1041 75
XM_017016645.1 XP_016872134.1 3 - 4 160 75
XM_017016647.1 XP_016872136.1 8 - 9 1041 75
XM_017016650.1 XP_016872139.1 8 - 9 1041 75
XM_017016651.1 XP_016872140.1 8 - 9 1041 75
GenBank mRNA AK057324.1 - 8 - 9 1041 75
BC022020.2 - 8 - 9 1239 75
BC042672.1 - 8 - 9 1024 75
BX647771.1 - 7 - 8 862 75
EF656619.1 - 6 - 7 587 75

View Full Product Details