Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL6
Entrez Gene ID
89857
Gene Name
kelch like family member 6
Gene Aliases
-
Chromosome Location
Chr.3: 183487531 - 183555712 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130446.2 NP_569713.2 3 - 4 963 66
XM_011513273.2 XP_011511575.1 2 - 3 618 66
GenBank mRNA AF441792.1 - 3 - 4 875 66
AK097125.1 - 3 - 4 917 66
AK314473.1 - 3 - 4 917 66
BC032348.1 - 3 - 4 941 66

View Full Product Details