Product Details
- Assay Reference Genome
Location
Chr.22:26452397 on build GRCh38
- Cytoband
- 22q12.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 89781
- Gene Symbol
- HPS4
- Gene Name
- HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
- Gene Aliases
- BLOC3S2, LE
- Location
- Chr.22:26443613-26483863 on build GRCh38
- Assay Gene Location
- Within Exon 17
View Full Product Details