Product Details

Assay Reference Genome
Location

Chr.1:109608640 on build GRCh38
Cytoband
1p13.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2780
Gene Symbol
GNAT2
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
Location
Chr.1:109603091-109619733 on build GRCh38
Assay Gene Location
Overlaps Intron 5 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GNAT2 NM_005272.3 NP_005263.1
XM_011541264.2 XP_011539566.1
AF493909.1 AAM12623.1
BC000233.1 AAH00233.1
BT009844.1 AAP88846.1

View Full Product Details