Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP14
Entrez Gene ID
57677
Gene Name
ZFP14 zinc finger protein
Gene Aliases
ZNF531
Chromosome Location
Chr.19: 36334453 - 36379203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297619.1 NP_001284548.1 1 - 2 42 61
NM_020917.2 NP_065968.1 1 - 2 42 61
GenBank mRNA AK293103.1 - 1 - 2 38 61
AL699477.1 - 1 - 2 43 61

View Full Product Details