The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080543.1 | NP_001074012.1 | 6 - 7 | 1210 | 90 |
NM_021231.1 | NP_067054.1 | 6 - 7 | 1210 | 90 | |
XM_011528160.2 | XP_011526462.1 | 6 - 7 | 1205 | 90 | |
GenBank mRNA | AK299963.1 | - | - | 1137 | 90 |
AY917150.1 | - | 6 - 7 | 1212 | 90 | |
CF272634.1 | - | 3 - 4 | 317 | 90 |