Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080423.3 NP_001073892.3 18 - 19 2258 65
XM_011534141.2 XP_011532443.1 18 - 19 2340 65
XM_011534142.2 XP_011532444.1 18 - 19 2252 65
XM_011534143.2 XP_011532445.1 17 - 18 2214 65
XM_011534145.2 XP_011532447.1 12 - 13 1986 65
XM_011534146.2 XP_011532448.1 17 - 18 2819 65
GenBank mRNA AB051506.2 - 18 - 19 2244 65
AK024507.1 - 5 - 6 598 65

View Full Product Details