The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099666.1 | NP_001093136.1 | 1 - 2 | 111 | 66 |
XM_005251976.4 | XP_005252033.1 | 1 - 2 | 201 | 66 | |
XM_005251977.4 | XP_005252034.1 | 1 - 2 | 175 | 66 | |
XM_005251980.4 | XP_005252037.1 | 1 - 2 | 161 | 66 | |
XM_005251981.4 | XP_005252038.1 | 1 - 2 | 161 | 66 | |
XM_011518640.1 | XP_011516942.1 | 1 - 2 | 195 | 66 | |
GenBank mRNA | BC053622.1 | - | 1 - 2 | 93 | 66 |