Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFPL3S
Entrez Gene ID
10737
Gene Name
RFPL3 antisense
Gene Aliases
NCRNA00005, RFPL3-AS, RFPL3-AS1, RFPL3ANT, RFPL3AS
Chromosome Location
Chr.22: 32359906 - 32371264 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_001450.3 3 - 4 278 90
NR_104232.1 4 - 5 210 90
GenBank mRNA AJ010233.1 - 3 - 4 187 90


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY010615.1 - 3 - 4 205 Chr. 22 - 32360508 - 32371002 [-] on Build GRCh38 90
GenBankmRNA HY040615.1 - 3 - 4 205 Chr. 22 - 32360579 - 32371003 [-] on Build GRCh38 90
GenBankmRNA HY027374.1 - 3 - 4 205 Chr. 22 - 32360612 - 32371002 [-] on Build GRCh38 90
GenBankmRNA HY035024.1 - 3 - 4 234 Chr. 22 - 32360541 - 32371031 [-] on Build GRCh38 90
GenBankmRNA BX117543.1 - 3 - 4 187 Chr. 22 - 32360271 - 32370985 [-] on Build GRCh38 90
GenBankmRNA BG185333.1 - 4 - 5 250 Chr. 22 - 32360761 - 32381253 [-] on Build GRCh38 90
GenBankmRNA HY030434.1 - 3 - 4 205 Chr. 22 - 32360589 - 32371003 [-] on Build GRCh38 90
GenBankmRNA BG198113.1 - 4 - 5 250 Chr. 22 - 32360219 - 32381253 [-] on Build GRCh38 90
GenBankmRNA HY034267.1 - 3 - 4 205 Chr. 22 - 32360596 - 32371002 [-] on Build GRCh38 90
GenBankmRNA HY028203.1 - 3 - 4 205 Chr. 22 - 32360565 - 32371002 [-] on Build GRCh38 90
GenBankmRNA HY004848.1 - 3 - 4 205 Chr. 22 - 32360571 - 32371002 [-] on Build GRCh38 90
GenBankmRNA HY002386.1 - 3 - 4 205 Chr. 22 - 32360553 - 32371003 [-] on Build GRCh38 90


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ret finger protein like 3 RFPL3 Anti-sense Chr. - 32354885 - 32361161 on Build GRCh38
Downstream uncharacterized LOC339666 LOC339666 9.0kb Anti-sense Chr. - 32376664 - 32384343 on Build GRCh38

View Full Product Details