Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MROH3P
- Entrez Gene ID
-
647215
- Gene Name
- maestro heat like repeat family member 3, pseudogene
- Gene Aliases
-
C1orf81
- Chromosome Location
-
Chr.1: 200917064 - 200966676 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
DQ983818.1 |
- |
2 - 3 |
205 |
127 |
View Full Product Details