The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128591.1 | NP_001122063.1 | 1 - 2 | 265 | 71 |
NM_001128592.1 | NP_001122064.1 | 1 - 2 | 265 | 71 | |
NM_001135750.1 | NP_001129222.1 | 1 - 2 | 265 | 71 | |
XM_011514593.2 | XP_011512895.1 | 1 - 2 | 260 | 71 | |
XM_011514594.2 | XP_011512896.1 | 1 - 2 | 260 | 71 | |
GenBank mRNA | AK126623.1 | - | 1 - 2 | 220 | 71 |
BM738331.1 | - | 1 - 2 | 242 | 71 | |
CN429483.1 | - | 1 - 2 | 233 | 71 | |
DA093618.1 | - | 1 - 2 | 220 | 71 |