Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM82
Entrez Gene ID
388595
Gene Name
transmembrane protein 82
Gene Aliases
-
Chromosome Location
Chr.1: 15742492 - 15747797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013641.2 NP_001013663.1 2 - 3 232 74
GenBank mRNA BC127706.1 - 2 - 3 212 74
BC127707.1 - 2 - 3 212 74
BC137239.1 - 2 - 3 232 74
BC137240.1 - 2 - 3 232 74

View Full Product Details