The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013641.2 | NP_001013663.1 | 4 - 5 | 828 | 110 |
GenBank mRNA | BC127706.1 | - | 4 - 5 | 808 | 110 |
BC127707.1 | - | 4 - 5 | 808 | 110 | |
BC137239.1 | - | 4 - 5 | 828 | 110 | |
BC137240.1 | - | 4 - 5 | 828 | 110 |