The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142641.1 | NP_001136113.1 | 12 - 13 | 1830 | 79 |
XM_005266171.3 | XP_005266228.1 | 15 - 16 | 1935 | 79 | |
XM_005266173.3 | XP_005266230.1 | 15 - 16 | 1930 | 79 | |
XM_005266175.3 | XP_005266232.1 | 14 - 15 | 1869 | 79 | |
XM_005266176.3 | XP_005266233.1 | 14 - 15 | 1811 | 79 | |
XM_005266177.3 | XP_005266234.1 | 15 - 16 | 1930 | 79 | |
XM_011534803.2 | XP_011533105.1 | 15 - 16 | 1994 | 79 | |
XM_011534804.2 | XP_011533106.1 | 15 - 16 | 1991 | 79 | |
XM_011534805.2 | XP_011533107.1 | 14 - 15 | 1925 | 79 | |
XM_011534806.2 | XP_011533108.1 | 14 - 15 | 1928 | 79 | |
XM_011534807.2 | XP_011533109.1 | 14 - 15 | 1870 | 79 | |
XM_011534808.2 | XP_011533110.1 | 15 - 16 | 1989 | 79 | |
XM_011534809.2 | XP_011533111.1 | 14 - 15 | 1806 | 79 | |
XM_011534810.2 | XP_011533112.1 | 15 - 16 | 1994 | 79 | |
XM_011534812.2 | XP_011533114.1 | 16 - 17 | 1816 | 79 | |
XM_011534816.2 | XP_011533118.1 | 15 - 16 | 1993 | 79 | |
GenBank mRNA | AB046765.2 | - | 12 - 13 | 1601 | 79 |
AF490258.1 | - | 5 - 6 | 696 | 79 |