The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005254670.2 | XP_005254727.1 | 1 - 2 | 88 | 81 |
XM_005254671.2 | XP_005254728.1 | 1 - 2 | 88 | 81 | |
XM_005254673.2 | XP_005254730.1 | 1 - 2 | 91 | 81 | |
XM_011522033.2 | XP_011520335.1 | 1 - 2 | 88 | 81 | |
XM_011522034.2 | XP_011520336.1 | 1 - 2 | 88 | 81 | |
XM_011522035.2 | XP_011520337.1 | 1 - 2 | 88 | 81 | |
XM_011522036.2 | XP_011520338.1 | 1 - 2 | 88 | 81 | |
XM_011522037.2 | XP_011520339.1 | 1 - 2 | 88 | 81 | |
XM_011522038.2 | XP_011520340.1 | 1 - 2 | 88 | 81 | |
XM_011522039.2 | XP_011520341.1 | 1 - 2 | 88 | 81 | |
XM_011522040.1 | XP_011520342.1 | 1 - 2 | 88 | 81 | |
XM_011522041.1 | XP_011520343.1 | 1 - 2 | 88 | 81 |