Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS6
Entrez Gene ID
253782
Gene Name
ceramide synthase 6
Gene Aliases
CERS5, LASS6
Chromosome Location
Chr.2: 168456249 - 168775134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256126.1 NP_001243055.1 1 - 2 364 67
NM_203463.2 NP_982288.1 1 - 2 364 67
GenBank mRNA AI521447.1 - 1 - 2 288 67
AK172775.1 - 1 - 2 364 67
BC109284.1 - 1 - 2 214 67
BC109285.1 - 1 - 2 214 67
BX393696.2 - 1 - 2 294 67

View Full Product Details