The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032283.2 | NP_115659.1 | 2 - 3 | 589 | 55 |
GenBank mRNA | AK056427.1 | - | 2 - 3 | 341 | 55 |
AK298683.1 | - | 2 - 3 | 196 | 55 | |
AL512765.1 | - | 2 - 3 | 194 | 55 | |
BC066776.1 | - | 2 - 3 | 589 | 55 |