The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081492.1 | NP_001074961.1 | 1 - 2 | 395 | 98 |
NM_182507.2 | NP_872313.2 | 1 - 2 | 395 | 98 | |
GenBank mRNA | AJ717743.1 | - | 1 - 2 | 349 | 98 |
BC065180.1 | - | 1 - 2 | 395 | 98 |