The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173351.1 | NP_775486.1 | 1 - 1 | 957 | 67 |
XM_011510671.2 | XP_011508973.1 | 3 - 3 | 1354 | 67 |