The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_135011.1 | 17 - 18 | 2586 | 118 | |
NR_135012.1 | 16 - 17 | 2079 | 118 | ||
GenBank mRNA | JX431285.1 | - | 16 - 17 | 1885 | 118 |
JX431286.1 | - | 17 - 18 | 2392 | 118 |