Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1LC3C
Entrez Gene ID
440738
Gene Name
microtubule associated protein 1 light chain 3 gamma
Gene Aliases
ATG8J, LC3C
Chromosome Location
Chr.1: 241995194 - 242001393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004343.2 NP_001004343.1 2 - 3 192 106
XM_005273139.3 XP_005273196.1 3 - 4 220 106
GenBank mRNA AF276659.1 - 2 - 3 181 106
BC127722.1 - 2 - 3 183 106
BC132986.1 - 2 - 3 172 106
BC132988.1 - 2 - 3 172 106
CN260933.1 - 2 - 3 192 106

View Full Product Details